(4-(4-Propylcyclohexyl)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

A161308-10G A161308-25G
77247-656EA 58.71 USD
77247-656 77247-658
(4-(4-Propylcyclohexyl)phenyl)boronic acid 98%
(4-(4-Propylcyclohexyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₅H₂₃BO₂
MW: 246.15 g/mol
MDL Number: MFCD00808678
CAS Number: 156837-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR