2-Fluoro-(5-methoxycarbonyl)phenylboronic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Fluoro-5-(methoxycarbonyl)phenylboronic acid, (2-Fluoro-5-(methoxycarbonyl)phenyl)boronic acid
A161255-100G
A161255-10G
A161255-1G
A161255-250MG
A161255-25G
A161255-5G
77235-860EA
897
USD
77235-860
77235-862
77235-864
77235-866
77235-868
77235-870
2-Fluoro-(5-methoxycarbonyl)phenylboronic acid 95%
2-Fluoro-(5-methoxycarbonyl)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₈H₈BFO₄ MW: 197.9600 g/mol |
MDL Number:
MFCD06659868 CAS Number: 850568-04-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...