6-Aminopyrimidine-4-carboxylic hemisulfuric acid salt 97%

Supplier: Ambeed
Warning

A161173-1G A161173-250MG A161173-5G
77273-308EA 56.65 USD
77273-308 77273-310 77273-312
6-Aminopyrimidine-4-carboxylic hemisulfuric acid salt 97%
6-Aminopyrimidine-4-carboxylic hemisulfuric acid salt

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₆O₈S
MW: 376.30 g/mol
MDL Number: MFCD31619106
CAS Number: 2173116-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR