3-Carboxy-4-methoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Borono-2-methoxybenzoic acid, 3-Carboxy-4-methoxybenzeneboronic acid

A161088-1G A161088-250MG A161088-5G
77273-296EA 15.45 USD
77273-296 77273-298 77273-300
3-Carboxy-4-methoxyphenylboronic acid 98%
3-Carboxy-4-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₅
MW: 195.97 g/mol
MDL Number: MFCD06203322
CAS Number: 913836-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR