3'-Fluoropropiophenone 97%

Supplier: Ambeed
Warning

Synonyms: m-Fluoropropiophenone, 1-(3-Fluorophenyl)propan-1-one

A160974-100G A160974-10G A160974-1G A160974-25G A160974-500G A160974-5G
77235-848EA 74.16 USD
77235-848 77235-850 77235-852 77235-854 77235-856 77235-858
3'-Fluoropropiophenone 97%
3'-Fluoropropiophenone

Precaución: for Research Use Only

Formula: C₉H₉FO
MW: 152.17 g/mol
MDL Number: MFCD00061245
CAS Number: 455-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR