Methyl-3-chloro-4-methoxybenzoate 95%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-methoxybenzoic acid methyl ester, Methyl 3-chloro-4-methoxybenzoate

A160853-1G A160853-5G
77273-288EA 6 USD
77273-288 77615-312
Methyl-3-chloro-4-methoxybenzoate 95%
Methyl-3-chloro-4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
MDL Number: MFCD00017188
CAS Number: 37908-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR