1,2-Dichloro-4-fluoro-5-nitrobenzene 98%

Supplier: Ambeed
Warning

A160733-10G A160733-1G A160733-5G
77286-428EA 21 USD
77286-428 77286-430 77286-432
1,2-Dichloro-4-fluoro-5-nitrobenzene 98%
1,2-Dichloro-4-fluoro-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂Cl₂FNO₂
MDL Number: MFCD00075330
CAS Number: 2339-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR