4-Morpholin-4-yl-1,2,5-thiadiazol-3-ol 95%

Supplier: Ambeed
Warning

Synonyms: 4-Morpholino-1,2,5-thiadiazol-3-ol

A160617-1G A160617-5G
77286-424EA 169.95 USD
77286-424 77286-426
4-Morpholin-4-yl-1,2,5-thiadiazol-3-ol 95%
4-Morpholin-4-yl-1,2,5-thiadiazol-3-ol

Precaución: for Research Use Only

Formula: C₆H₉N₃O₂S
MW: 187.22 g/mol
MDL Number: MFCD01820806
CAS Number: 30165-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR