(3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methanol 97%

Supplier: Ambeed
Warning

A160493-100G A160493-10G A160493-1G A160493-25G A160493-5G
77259-598EA 157.59 USD
77259-598 77259-600 77259-602 77259-604 77259-606
(3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methanol 97%
(3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₀F₃NO₂
MW: 221.17 g/mol
MDL Number: MFCD07368269
CAS Number: 103577-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR