2-Fluoro-5-methylpyridine 97%

Supplier: Ambeed
Danger

Synonyms: 2-Fluoro-5-picoline

A160424-10G A160424-1G A160424-5G A160424-25G
77273-274EA 11.33 USD
77273-274 77273-276 77273-278 77592-954
2-Fluoro-5-methylpyridine 97%
2-Fluoro-5-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₆FN
MW: 111.12 g/mol
MDL Number: MFCD00041225
CAS Number: 2369-19-9
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR