6-Bromoisoquinoline 98%

Supplier: Ambeed

A160132-100G A160132-10G A160132-1G A160132-25G A160132-500G A160132-5G
77259-578EA 186 USD
77259-578 77259-580 77259-582 77259-584 77259-586 77259-588
6-Bromoisoquinoline 98%
6-Bromoisoquinoline

Precaución: for Research Use Only

Formula: C₉H₆BrN
MDL Number: MFCD04973299
CAS Number: 34784-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR