1-((1H-Indol-4-yl)oxy)-3-(tert-butyl(4-methylbenzyl)amino)propan-2-ol 98%

Supplier: Ambeed
Warning

A160003-10MG A160003-1MG A160003-25MG A160003-50MG A160003-5MG
77259-552EA 97.85 USD
77259-552 77259-554 77259-556 77259-558 77259-560
1-((1H-Indol-4-yl)oxy)-3-(tert-butyl(4-methylbenzyl)amino)propan-2-ol 98%
1-((1H-Indol-4-yl)oxy)-3-(tert-butyl(4-methylbenzyl)amino)propan-2-ol

Precaución: for Research Use Only

Formula: C₂₃H₃₀N₂O₂
MW: 366.49 g/mol
MDL Number: MFCD31657330
CAS Number: 2095432-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR