2,5-Dibromoadipic acid 97%

Supplier: Ambeed
Danger

Synonyms: 2,5-Dibromohexanedioic acid

A159987-1G A159987-250MG A159987-25G A159987-5G
77247-638EA 71.07 USD
77247-638 77247-640 77247-642 77247-644
2,5-Dibromoadipic acid 97%
2,5-Dibromoadipic acid

Precaución: for Research Use Only

Formula: C₆H₈Br₂O₄
MW: 303.95 g/mol
MDL Number: MFCD00046516
CAS Number: 3479-85-4
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR