3-(tert-Butyl)-9H-carbazole 95%

Supplier: Ambeed
Warning

A159978-100MG A159978-1G A159978-250MG
77247-632EA 14.42 USD
77247-632 77247-634 77247-636
3-(tert-Butyl)-9H-carbazole 95%
3-(tert-Butyl)-9H-carbazole

Precaución: for Research Use Only

Formula: C₁₆H₁₇N
MW: 223.32 g/mol
MDL Number: MFCD12024281
CAS Number: 22401-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR