2,6-Dichloro-5-methoxynicotinonitrile 97%

Supplier: Ambeed
Warning

A159929-1G A159929-250MG
77259-542EA 309 USD
77259-542 77259-544
2,6-Dichloro-5-methoxynicotinonitrile 97%
2,6-Dichloro-5-methoxynicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₄Cl₂N₂O
MW: 203.03 g/mol
MDL Number: MFCD30730107
CAS Number: 2089381-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR