Isoluminol (4-aminophthalhydrazide) 98%

Supplier: Ambeed
Warning

Synonyms: 6-Amino-2,3-dihydro-1,4-phthalazinedione, 4-Aminophthaloylhydrazine, 6-Amino-2,3-dihydrophthalazine-1,4-dione, Isoluminol, 4-Aminophthalhydrazide

A159775-250MG A159775-1G A159775-25G A159775-5G
77235-810EA 7.21 USD
77235-810 77615-302 77746-091 77746-092
Isoluminol (4-aminophthalhydrazide) 98%
Isoluminol (4-aminophthalhydrazide)

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
MW: 177.16 g/mol
MDL Number: MFCD00010560
CAS Number: 3682-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR