(2,3,5,6-Tetrafluorophenoxy)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(2,3,5,6-Tetrafluorophenoxy)acetic acid

A159587-100MG A159587-250MG A159587-5G
77247-616EA 28.84 USD
77247-616 77247-618 77247-620
(2,3,5,6-Tetrafluorophenoxy)acetic acid 97%
(2,3,5,6-Tetrafluorophenoxy)acetic acid

Precaución: for Research Use Only

Formula: C₈H₄F₄O₃
MW: 224.11 g/mol
MDL Number: MFCD01166305
CAS Number: 14742-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR