2-(Chloromethyl)benzothiazole 98%

Supplier: Ambeed
Danger

Synonyms: 2-(Chloromethyl)-1,3-benzothiazole

A159515-1G A159515-5G A159515-10G
77235-800EA 13.39 USD
77235-800 77550-004 77592-942 77746-088
2-(Chloromethyl)benzothiazole 98%
2-(Chloromethyl)benzothiazole

Precaución: for Research Use Only

Formula: C₈H₆CINS
MDL Number: MFCD05664944
CAS Number: 37859-43-1
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR