2-Amino-5-methoxy-1-methylbenzimidazole 97%

Supplier: Ambeed
Warning

A159478-100MG A159478-1G A159478-250MG
77247-610EA 20 USD
77247-610 77247-612 77247-614
2-Amino-5-methoxy-1-methylbenzimidazole 97%
2-Amino-5-methoxy-1-methylbenzimidazole

Precaución: for Research Use Only

Formula: C₉H₁₁N₃O
MDL Number: MFCD01658296
CAS Number: 1805-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR