3,4-Dimethoxycinnamic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dimethoxycinnamic Acid

A159365-100G A159365-10G A159365-25G A159365-5G
77273-246EA 47.38 USD
77273-246 77273-248 77319-348 77615-294
3,4-Dimethoxycinnamic acid 98%
3,4-Dimethoxycinnamic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₄
MW: 208.21 g/mol
MDL Number: MFCD00004387
CAS Number: 2316-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR