2,3-Difluoro-4-iodopyridine 98%

Supplier: Ambeed
Warning

A159196-100MG A159196-10G A159196-1G A159196-250MG A159196-25G A159196-5G
77247-598EA 5.15 USD
77247-598 77247-600 77247-602 77247-604 77247-606 77247-608
2,3-Difluoro-4-iodopyridine 98%
2,3-Difluoro-4-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₂F₂IN
MW: 240.9800 g/mol
MDL Number: MFCD09909609
CAS Number: 851386-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR