MK-4827 (Tosylate) ≥98%

Supplier: Ambeed
Danger

Synonyms: MK-4827 Tosylate, Niraparib tosylate, (3S)-3-[4-[7-(Aminocarbonyl)-2H-indazol-2-yl]phenyl]piperidine Tosylate, (S)-2-(4-(piperidin-3-yl)phenyl)-2H-indazole-7-carboxamide 4-methylbenzenesulphonate

A159121-1MG A159121-10MG A159121-2MG A159121-5MG
77235-788EA 10.3 USD
77235-788 77746-084 77746-085 77746-086
MK-4827 (Tosylate) ≥98%
MK-4827 (Tosylate)

Precaución: for Research Use Only

Formula: C₁₉H₂₀N₄O·C₇H₈O₃S
MW: 492.6 g/mol
MDL Number: MFCD28167748
CAS Number: 1038915-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR