2-Chloro-4-phenylpyridine 97%

Supplier: Ambeed
Warning

A158968-100MG A158968-25G A158968-5G
77259-508EA 11.33 USD
77259-508 77259-510 77259-512
2-Chloro-4-phenylpyridine 97%
2-Chloro-4-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₁H₈ClN
MW: 189.6400 g/mol
MDL Number: MFCD04114163
CAS Number: 42260-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR