2,4-Di-tert-pentylphenol 99%

Supplier: Ambeed
Warning

A158947-100G A158947-25G
77247-584EA 21 USD
77247-584 77247-586
2,4-Di-tert-pentylphenol 99%
2,4-Di-tert-pentylphenol

Precaución: for Research Use Only

Formula: C₁₆H₂₆O
MDL Number: MFCD00041929
CAS Number: 120-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR