4-Phenyl-1-buten-4-ol 98%

Supplier: Ambeed
Warning

Synonyms: 1-Phenyl-3-buten-1-ol

A158843-1G A158843-5G
77235-776EA 59.74 USD
77235-776 77235-778
4-Phenyl-1-buten-4-ol 98%
4-Phenyl-1-buten-4-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
MW: 148.20 g/mol
MDL Number: MFCD00039617
CAS Number: 936-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR