6-Hydroxy-2-methylisoindolin-1-one 97%

Supplier: Ambeed
Warning

A158831-100MG A158831-1G A158831-250MG
77235-770EA 37 USD
77235-770 77235-772 77235-774
6-Hydroxy-2-methylisoindolin-1-one 97%
6-Hydroxy-2-methylisoindolin-1-one

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MW: 163.17 g/mol
MDL Number: MFCD20441063
CAS Number: 1344701-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR