(S)-(-)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol ≥97%, ee 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-[1,1'-binaphthalene]-2,2'-diol
A158819-1G
A158819-250MG
A158819-5G
77247-578EA
39
USD
77247-578
77247-580
77247-582
(S)-(-)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol ≥97%, ee 99%
(S)-(-)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol
Precaución: for Research Use Only
Formula:
C₂₀H₂₀Br₂O₂ MW: 452.1800 g/mol |
MDL Number:
MFCD03093977 CAS Number: 765278-73-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...