2-(4-(2-Fluoro-5-(3-(6-methylpyridin-2-yl)-1H-pyrazol-4-yl)phenyl)-1H-pyrazol-1-yl)ethan-1-ol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
R-268712
A158772-10MG
A158772-1MG
A158772-25MG
A158772-50MG
A158772-5MG
77235-760EA
50.47
USD
77235-760
77235-762
77235-764
77235-766
77235-768
2-(4-(2-Fluoro-5-(3-(6-methylpyridin-2-yl)-1H-pyrazol-4-yl)phenyl)-1H-pyrazol-1-yl)ethan-1-ol 98%
2-(4-(2-Fluoro-5-(3-(6-methylpyridin-2-yl)-1H-pyrazol-4-yl)phenyl)-1H-pyrazol-1-yl)ethan-1-ol
Precaución: for Research Use Only
Formula:
C₂₀H₁₈FN₅O MW: 363.38 g/mol |
MDL Number:
MFCD28963939 CAS Number: 879487-87-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...