2',3,4,5-Tetrafluoro-4'-(trans-4-propylcyclohexyl)biphenyl 97%

Supplier: Ambeed
Warning

Synonyms: rel-2,3',4',5'-Tetrafluoro-4-((1s,4r)-4-propylcyclohexyl)-1,1'-biphenyl

A158668-100G A158668-1G A158668-25G A158668-5G
77247-570EA 445 USD
77247-570 77247-572 77247-574 77247-576
2',3,4,5-Tetrafluoro-4'-(trans-4-propylcyclohexyl)biphenyl 97%
2',3,4,5-Tetrafluoro-4'-(trans-4-propylcyclohexyl)biphenyl

Precaución: for Research Use Only

Formula: C₂₁H₂₂F₄
MW: 350.40 g/mol
MDL Number: MFCD22380673
CAS Number: 173837-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR