1H-Pyrazole-3-carbonitrile 98%

Supplier: Ambeed
Warning

A158600-100MG A158600-1G A158600-250MG A158600-5G
77235-738EA 11.33 USD
77235-738 77235-740 77235-742 77235-744
1H-Pyrazole-3-carbonitrile 98%
1H-Pyrazole-3-carbonitrile

Precaución: for Research Use Only

Formula: C₄H₃N₃
MW: 93.0900 g/mol
MDL Number: MFCD08669496
CAS Number: 36650-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR