β-L-Thymidine 98%

Supplier: Ambeed
Warning

Synonyms: Telbivudine, 1-((2S,4R,5S)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione

A158594-10MG A158594-1G A158594-250MG A158594-25G
77259-502EA 5.15 USD
77259-502 77259-504 77259-506 77748-073
β-L-Thymidine 98%
β-L-Thymidine

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₅
MW: 242.2286 g/mol
MDL Number: MFCD02683612
CAS Number: 3424-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR