H-Orn(Z)-OBzl·HCl 98%

Supplier: Ambeed
Warning

Synonyms: (S)-Benzyl 2-amino-5-(((benzyloxy)carbonyl)amino)pentanoate hydrochloride, H-Orn(Z)-OBzl·HCl

A158547-1G A158547-5G
77235-734EA 23.69 USD
77235-734 77235-736
H-Orn(Z)-OBzl·HCl 98%
H-Orn(Z)-OBzl·HCl

Precaución: for Research Use Only

Formula: C₂₀H₂₅ClN₂O₄
MW: 392.88 g/mol
MDL Number: MFCD00237210
CAS Number: 63594-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR