5-(Benzyloxy)-2-(hydroxymethyl)pyridin-4(1H)-one 97%

Supplier: Ambeed
Warning

A158391-1G A158391-10G
77247-566EA 47 USD
77247-566 77567-300
5-(Benzyloxy)-2-(hydroxymethyl)pyridin-4(1H)-one 97%
5-(Benzyloxy)-2-(hydroxymethyl)pyridin-4(1H)-one

Precaución: For research use only.

Formula: C₁₃H₁₃NO₃
MDL Number: MFCD00100501
CAS Number: 59281-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR