6-Bromo-8-methylimidazo[1,2-a]pyrazine 97%

Supplier: Ambeed
Warning

A158346-100MG A158346-250MG
77235-730EA 21 USD
77235-730 77235-732
6-Bromo-8-methylimidazo[1,2-a]pyrazine 97%
6-Bromo-8-methylimidazo[1,2-a]pyrazine

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MDL Number: MFCD12033249
CAS Number: 1159815-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR