5-Methyl-4H-1,2,4-triazol-3-amine 98%

Supplier: Ambeed
Warning

A158293-10G A158293-1G A158293-250MG A158293-25G A158293-5G
77273-204EA 105 USD
77273-204 77273-206 77273-208 77273-210 77273-212
5-Methyl-4H-1,2,4-triazol-3-amine 98%
5-Methyl-4H-1,2,4-triazol-3-amine

Precaución: for Research Use Only

Formula: C3H6N4
MDL Number: MFCD00498236
CAS Number: 4923-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR