3-Fluorothiophene-2-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-2-thiophenecarboxylic acid

A158289-10G A158289-1G A158289-250MG A158289-5G
77273-196EA 347.11 USD
77273-196 77273-198 77273-200 77273-202
3-Fluorothiophene-2-carboxylic acid 98%
3-Fluorothiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C5H3FO2S
MW: 146.1395 g/mol
MDL Number: MFCD03426911
CAS Number: 32431-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR