(2-Bromo-4-chlorophenyl)methanamine hydrochloride 97%

Supplier: Ambeed
Warning

A158184-100MG A158184-1G A158184-250MG
77247-558EA 25.75 USD
77247-558 77247-560 77247-562
(2-Bromo-4-chlorophenyl)methanamine hydrochloride 97%
(2-Bromo-4-chlorophenyl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₈BrCl₂N
MW: 256.96 g/mol
MDL Number: MFCD30609557
CAS Number: 2048273-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR