1,3,5-Tris(4-formylphenyl)benzene 97%

Supplier: Ambeed
Warning

Synonyms: 5'-(4-Formylphenyl)-[1,1':3',1''-terphenyl]-4,4''-dicarboxaldehyde, 5'-(4-Formylphenyl)-[1,1':3',1''-terphenyl]-4,4''-dicarbaldehyde

A157674-1G A157674-250MG A157674-5G
77247-542EA 66.95 USD
77247-542 77247-544 77247-546
1,3,5-Tris(4-formylphenyl)benzene 97%
1,3,5-Tris(4-formylphenyl)benzene

Precaución: for Research Use Only

Formula: C₂₇H₁₈O₃
MW: 390.44 g/mol
MDL Number: MFCD27937712
CAS Number: 118688-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR