2-Amino-4,6-dihydroxypyrimidine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-6-hydroxy-1H-pyrimidin-4-one, 2-Amino-6-hydroxypyrimidin-4(3H)-one

A157653-1000G A157653-100G A157653-10G A157653-25G A157653-500G
77286-330EA 162 USD
77286-330 77286-332 77286-334 77286-336 77286-338
2-Amino-4,6-dihydroxypyrimidine 95%
2-Amino-4,6-dihydroxypyrimidine

Precaución: for Research Use Only

Formula: C₄H₅N₃O₂
MW: 127.1 g/mol
Boiling Pt: 394
Melting Pt: 300
MDL Number: MFCD00006094
CAS Number: 56-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR