Ethyl 2-((tetrahydro-2H-pyran-2-yl)oxy)acetate 95%

Supplier: Ambeed
Warning

A157586-1G A157586-250MG A157586-5G
77235-722EA 116.39 USD
77235-722 77235-724 77235-726
Ethyl 2-((tetrahydro-2H-pyran-2-yl)oxy)acetate 95%
Ethyl 2-((tetrahydro-2H-pyran-2-yl)oxy)acetate

Precaución: for Research Use Only

Formula: C₉H₁₆O₄
MW: 188.22 g/mol
MDL Number: MFCD00128492
CAS Number: 61675-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR