2-Bromo-5-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-5-picoline

A157488-100G A157488-10G A157488-500G A157488-5G A157488-25G
77273-174EA 54 USD
77273-174 77273-176 77273-178 77273-180 77319-038
2-Bromo-5-methylpyridine 98%
2-Bromo-5-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 65
Melting Pt: 152.5
MDL Number: MFCD00209553
CAS Number: 3510-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR