2'-Bromo-4'-chloroacetophenone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Bromo-4-chlorophenyl)ethanone

A157473-100G A157473-10G A157473-1G A157473-5G
77235-714EA 1273.08 USD
77235-714 77235-716 77235-718 77235-720
2'-Bromo-4'-chloroacetophenone 97%
2'-Bromo-4'-chloroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆BrClO
MW: 233.4896 g/mol
MDL Number: MFCD02683063
CAS Number: 825-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR