4-(2,3-Dichlorophenyl)-5-(methoxycarbonyl)-2,6-dimethyl-1,4-dihydropyridine-3-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A157376-100MG
A157376-1G
A157376-250MG
A157376-5G
77286-314EA
7
USD
77286-314
77286-316
77286-318
77286-320
4-(2,3-Dichlorophenyl)-5-(methoxycarbonyl)-2,6-dimethyl-1,4-dihydropyridine-3-carboxylic acid 95%
4-(2,3-Dichlorophenyl)-5-(methoxycarbonyl)-2,6-dimethyl-1,4-dihydropyridine-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₆H₁₅Cl₂NO₄ |
MDL Number:
MFCD06654363 CAS Number: 123853-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...