2,6-Difluoro-3-nitrobenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A157284-100G
A157284-10G
A157284-1G
A157284-25G
A157284-500G
77247-522EA
83
USD
77247-522
77247-524
77247-526
77247-528
77247-530
2,6-Difluoro-3-nitrobenzoic acid 98%
2,6-Difluoro-3-nitrobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₃F₂NO₄ |
MDL Number:
MFCD00119291 CAS Number: 83141-10-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...