3-Amino-2-fluorobenzamide 95%

Supplier: Ambeed
Warning

A156970-100MG A156970-250MG
77235-700EA 27 USD
77235-700 77235-702
3-Amino-2-fluorobenzamide 95%
3-Amino-2-fluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₇FN₂O
MDL Number: MFCD28722350
CAS Number: 1369948-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR