3,6-Dichloropyridazine-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A156968-1G A156968-25G A156968-5G A156968-10G
77273-158EA 14 USD
77273-158 77273-160 77273-162 77615-276
3,6-Dichloropyridazine-4-carboxylic acid 98%
3,6-Dichloropyridazine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₂Cl₂N₂O₂
MDL Number: MFCD00011575
CAS Number: 51149-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR