2-(Dimethylaminomethyl)-1-cyclohexanone hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 2-((Dimethylamino)methyl)cyclohexanone hydrochloride, Tramadol Related Compound B

A156911-10G A156911-1G A156911-25G A156911-5G A156911-100G
77247-514EA 39 USD
77247-514 77247-516 77247-518 77247-520 77567-280
2-(Dimethylaminomethyl)-1-cyclohexanone hydrochloride 97%
2-(Dimethylaminomethyl)-1-cyclohexanone hydrochloride

Precaución: For research use only.

Formula: C₉H₁₈ClNO
MDL Number: MFCD00102580
CAS Number: 42036-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR