4-Methoxypyridine-2,6-dicarboxylic acid 95%

Supplier: Ambeed
Warning

A156829-1G A156829-250MG A156829-5G
77247-508EA 187.46 USD
77247-508 77247-510 77247-512
4-Methoxypyridine-2,6-dicarboxylic acid 95%
4-Methoxypyridine-2,6-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₅
MW: 197.15 g/mol
MDL Number: MFCD11036211
CAS Number: 52062-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR