2,3-Dimethoxybenzylamine 95%

Supplier: Ambeed
Warning

Synonyms: 1-(2,3-Dimethoxyphenyl)methanamine, (2,3-Dimethoxyphenyl)methanamine

A156744-10G A156744-1G A156744-25G A156744-5G
77247-500EA 117.42 USD
77247-500 77247-502 77247-504 77247-506
2,3-Dimethoxybenzylamine 95%
2,3-Dimethoxybenzylamine

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂
MW: 167.21 g/mol
MDL Number: MFCD00052392
CAS Number: 4393-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR