(3R,7aR)-3-(tert-Butyl)-1,5-dioxohexahydropyrrolo[1,2-c]oxazole-7a-carbaldehyde 97%
Supplier: Ambeed
|
Warning
|
A156667-1G
A156667-250MG
77259-482EA
92.7
USD
77259-482
77259-484
(3R,7aR)-3-(tert-Butyl)-1,5-dioxohexahydropyrrolo[1,2-c]oxazole-7a-carbaldehyde 97%
(3R,7aR)-3-(tert-Butyl)-1,5-dioxohexahydropyrrolo[1,2-c]oxazole-7a-carbaldehyde
Precaución: for Research Use Only
Formula:
C₁₁H₁₅NO₄ MW: 225.24 g/mol |
MDL Number:
MFCD20488025 CAS Number: 1214741-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...